Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Epiregulin Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | Epiregulin |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119096
|
Invitrogen™
PA5119096 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122013
|
Invitrogen™
PA5122013 |
20 μL |
Each for $178.40
|
|
|||||
Description
Epiregulin Polyclonal Antibody for Western Blot
Epiregulin is an EGF-related growth factor that binds specifically to EGFR (ErbB1) and ErbB4, but not ErbB2 or ErbB3. It is expressed mainly in the placenta and peripheral blood leukocytes, as well as in certain carcinomas of the bladder, lung, kidney and colon. Epiregulin stimulates the proliferation of keratinocytes, hepatocytes, fibroblasts and vascular smooth muscle cells. It also inhibits the growth of several tumor-derived epithelial cell lines. Human Epiregulin is initially synthesized as a glycosylated 19.0 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a 6.0 kDa mature secreted sequence.Specifications
Epiregulin | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Ereg | |
Ep; Epiregulin; EPR; ER; EREG; Proepiregulin | |
Ereg | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O14944 | |
2069 | |
Recombinant protein Epiregulin. The antigen corresponds to amino acid range 30-169 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title