Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epirubicin HydroChloride, PROMO, Thermo Scientific Chemicals

Catalog No. J60411MAPM
Click to view available options
Quantity:
10 mg

Cell-permeable anthracycline anti-tumor antibiotic

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56390-09-1
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98 g/mol
MDL Number MFCD00941448
InChI Key MWWSFMDVAYGXBV-DPZLWMTJSA-N
Synonym Epiadriamycin
PubChem CID 123134690
IUPAC Name (7S,9S)-7-[(2R,4R,5R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)CO)O)N)O.Cl

Specifications

Melting Point 185°C
Color Red
Quantity 10 mg
Merck Index 14,3623
Solubility Information Soluble in DMSO to 100 mM, or in ethanol to 10 mM
Physical Form Solid
Chemical Name or Material Epirubicin hydrochloride
EINECSNumber 260-145-2
RTECSNumber QI9295750
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.