Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Episomal iPSC Reprogramming Vectors

Catalog No. A14703
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14703 1 tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14703 Supplier Invitrogen™ Supplier No. A14703

Episomal iPSC Reprogramming Vectors

Episomal iPSC Reprogramming Vectors are a cost-effective mixture of three vectors designed to provide the optimal system for generating transgene-free and virus-free induced pluripotent stem cells (iPSCs) in a feeder-free environment. Originally developed by Junying Yu and James Thomson(1) and further optimized by Cellular Dynamics International, these Episomal iPSC Reprogramming Vectors have proven successful in reprogramming a number of different somatic cell types.
  • Safe for all stages of your iPSC research—transgene-free and viral-free reprogramming allows use from basic to pre-clinical research
  • Reprogram a variety of somatic cell types—provides flexibility in somatic cell selection
  • Optimized for feeder-free reprogramming—allows for defined and feeder-free reprogramming when used with Essential 8™ Medium

    Create Transgene- and Virus-free iPSCs
    The Episomal iPSC Reprogramming Vectors are a well-described system for producing transgene-free, virus-free iPSCs, providing a source of iPSCs for all stages of your pluripotent stem cell research. Other reprogramming methods, such as lentivirus, contain transgenes that can integrate into the host genome, potentially disrupting the genome or causing unpredictable results. These episomal vectors are introduced into the cell by electroporation. As oriP/EBNA1 vectors, they contain all 6 reprogramming factors (Oct4, Sox2, Nanog, Lin28, Klf4 and lMyc) and replicate extrachromosomally only once per cell cycle. At this replication rate, the episomes are lost at a rate of approximately 5% per cell generation.

    Generate iPSCs from a Wide Variety of Somatic Cell Types
    iPSCs have been generated with episomal vectors from a range of somatic cells including fibroblasts, bone marrow mononuclear cells, PBMCs, lymphoblast B cells, and various disease-type fibroblasts and PBMCs. Each kit provides enough material for 6 reprogramming experiments.

    Optimized for Feeder-free Reprogramming with Essential 8™ Medium
    The Episomal iPSC Reprogramming Vectors were designed in the laboratories of James Thomson and Cellular Dynamics International for use with Essential 8™ Medium, thus providing an optimal environment for defined, feeder-free reprogramming.

    Commercialized in Partnership with Cellular Dynamics International.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

    Reference:
    1. Human Induced Pluripotent Stem Cells Free of Vector and Transgene Sequences; Yu J, Hu K, Smuga-Otto K, Tian S, Stewart R, Slukvin II, Thomson J. Science. 2009; 324:797-801.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Constitutive or Inducible System Constitutive
    Delivery Type Transfection
    For Use With (Equipment) Electroporator, Neon™ Transfection System
    Promoter CMV
    Product Type Episomal iPSC Reprogramming Vectors
    Selection Agent (Eukaryotic) Hygromycin
    Content And Storage 3 DNA vectors mixed into 1 tube at 1 μg/μl in TE buffer
    Replication Origin oriP
    Protein Tag Untagged
    Quantity 1 tube
    Vector pCEP4
    For Use With (Application) Episomal Expression, Induced Pluripotent Stem Cell Generation, Multi-Gene Expression
    Show More Show Less
    Which kits do you offer for stem cell reprogramming?

    We offer the Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit (Cat. Nos. A16517, A16518) which is a non-integrating system that uses Sendai virus vectors to reprogram somatic cells into induced pluripotent stem cells (iPSCs). The Invitrogen CytoTune-iPS 2.0 Sendai Reprogramming Kit contains three Invitrogen CytoTune 2.0 reprogramming vectors that are used for delivering and expressing key genetic factors necessary for reprogramming somatic cells into iPSCs. Only one application of the vectors is required for successful reprogramming.

    Additionally, we offer the Episomal iPSC Reprogramming Vectors (Cat. No A14703) or Epi5 Episomal iPSC Reprogramming Kit (Cat. No A15960). This is a non-integrating system that reprograms somatic cells into induced pluripotent stem cells (iPSCs). These two products contain a mixture of six or five vectors designed to provide the optimal system for generating transgene-free and virus-free iPSCs in a feeder-free environment. Originally developed by Junying Yu and James Thomson (http://www.ncbi.nlm.nih.gov/pubmed/19325077) and further optimized by Cellular Dynamics International, these Episomal iPSC Reprogramming Vectors have proven successful in reprogramming a number of different somatic cell types.

    When should I use the full length Human FGF-basic (FGF-2/bFGF) Recombinant Protein or the truncated variant, Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein?

    The full length Human FGF-basic (FGF-2/bFGF) (aa 1-155) Recombinant Protein is recommended for stem cells whereas the truncated variant, Human FGF-basic (FGF-2/bFGF) (aa 10-155) Recombinant Protein which is missing the first 9 amino acids, is recommended for use with neural and cardiac cells.

    What are induced pluripotent stem cells?

    Induced pluripotent stem cells (iPS or iPSCs) are pluripotent stem cells directly generated by introducing combination of genes coding for “reprogramming factors” into adult cells. These reprogramming factors include Oct4, Sox2, c-Myc, KLF4, NANOG, and LIN28. Yu, et al, generated iPS from a human mesenchymal cell line using lentiviral vectors carrying Oct4, Sox2, NANOG, and LIN28 genes (Science 318:1917 (2007)). Using a similar approach, Takahashi et al, generated iPS from human primary fibroblast cells by introducing genes coding for Oct3, Sox2, KLF4, and c-Myc into these cells (Cell 131:861 (2007)). iPS generated by reprogramming are similar to human ES cells in morphology, the capacity for unlimited proliferation, surface-antigen expression, gene expression, the ability to differentiate into cell types representing the three germ layers in vitro, and the ability to form teratomas after injection into SCID mice.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.