Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epithelial cell marker Antibody (LH39), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002690AF594
Description
Epithelial cell marker Monoclonal specifically detects Epithelial cell marker in Primate samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Epithelial cell marker | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
LH39 | |
Western Blot, Immunohistochemistry | |
Mouse | |
Protein A purified | |
Cancer, Cellular Markers, Extracellular Matrix | |
Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction