Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epithelial cell marker Antibody (LH39), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002690MFV610
Description
Epithelial cell marker Monoclonal antibody specifically detects Epithelial cell marker in Primate samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Epithelial cell marker | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Cells from a single cell suspension of epidermal cells (obtained from fresh human neonatal foreskin) were lysed in Nonidet P40 in phosphate buffered saline and the insoluble pellet was sonicated to prepare insoluble fractions. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LH39 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction