Learn More
Description
Host Species: Mouse
Clone: GP1.4
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: DTRP
Immunogen: Human milk fat globule membranes
Molecular Weight: 265-400 kDa
Positive Control: Breast or Colon CA
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Epithelial Membrane Antigen/EMA Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | GP1.4 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human milk fat globule membranes |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.