Learn More
epithelial Sodium Channel alpha Antibody (2G4), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259382FR
Catalog No. NBP259382FR
Only null left
Add to Cart
Description
Description
epithelial Sodium Channel alpha Monoclonal specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
epithelial Sodium Channel beta
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
SCNN1B
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68)
100 μL
Primary
Detects 85kDa.
Store at 4C in the dark.
IgG2b
Western Blot, Immunohistochemistry
2G4
Western Blot, Immunohistochemistry
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
Protein G purified
Phospho Specific
6338
Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.