Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB10074357
Description
epithelial Sodium Channel alpha Polyclonal specifically detects epithelial Sodium Channel alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
epithelial Sodium Channel alpha | |
Polyclonal | |
Unconjugated | |
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
SCNN1A | |
Synthetic peptide corresponding to residues L(20) M K G N K R E E Q G L G P E P A A P Q Q P T(42) C of Human alpha-Enac. | |
Affinity Purified | |
RUO | |
6337 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:100 - 1:1000, Flow Cytometry 3-5 ug/10^6 cells, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha | |
Rabbit | |
76 kDa | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction