Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel beta Antibody (16E4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259381AF488
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot.Specifications
epithelial Sodium Channel beta | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCNN1B | |
Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638) | |
100 μL | |
Primary | |
Detects 87kDa. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot | |
160000 | |
Western Blot | |
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
Mouse | |
Protein G purified | |
Phospho Specific | |
6338 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction