Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel beta Antibody (16E4), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | epithelial Sodium Channel beta |
---|---|
Clone | 160000 |
Dilution | Western Blot 1:100 - 1:2000 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot.Specifications
epithelial Sodium Channel beta | |
Western Blot 1:100 - 1:2000 | |
Unconjugated | |
Mouse | |
Phospho Specific | |
6338 | |
Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638) | |
Primary | |
Detects 87kDa. |
160000 | |
Monoclonal | |
Purified | |
RUO | |
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
SCNN1B | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title