Learn More
epithelial Sodium Channel beta Antibody (7B8), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259380H
Catalog No. NBP259380H
Only null left
Add to Cart
Description
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
epithelial Sodium Channel beta
Monoclonal
HRP
PBS with No Preservative
SCNN1B
Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638)
100 μL
Primary
Detects 87kDa.
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry
7B8
Western Blot, Immunohistochemistry
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
Protein G purified
Phospho Specific
6338
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.