Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel gamma Antibody (11-35-1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241374AF750
Description
epithelial Sodium Channel gamma Monoclonal specifically detects epithelial Sodium Channel gamma in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
epithelial Sodium Channel gamma | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SCNN1G | |
Synthetic peptide seguence: VGGSIIHKAC | |
Primary | |
Antibody is specific for the neoepitope generated after proteolytic cleavage of human gENaC and recognizes the C-terminal fragment starting from amino acid residue 182. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
11-35-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma | |
Mouse | |
0.1 mL | |
6340 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction