Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel gamma Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318256
This item is not returnable.
View return policy
Description
epithelial Sodium Channel gamma Polyclonal antibody specifically detects epithelial Sodium Channel gamma in Human, Mouse, Rat, Hamster, Xenopus samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
epithelial Sodium Channel gamma | |
Polyclonal | |
Unconjugated | |
PBS, 50% glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
6340 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
1 mg/ml | |
Western Blot 1:1000, Immunohistochemistry 1:100 | |
amiloride-sensitive epithelial sodium channel gamma subunit, amiloride-sensitive sodium channel gamma-subunit, BESC3, ENaC gamma subunit, ENaCG, ENaCgamma, Epithelial Na(+) channel subunit gamma, gamma-ENaC, gamma-NaCH, Nonvoltage-gated sodium channel 1 subunit gamma, PHA1, SCNEGamiloride-sensitive sodium channel subunit gamma, sodium channel, nonvoltage-gated 1, gamma | |
Produced against the C-terminal tail (amino acids 629-650) of rat gamma ENaC (antibody designation L550) | |
100 μg | |
Primary | |
Human, Mouse, Rat, Hamster, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction