Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS144P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS144P0 100 μL Unconjugated Concentrated 200μg/mL
MS144P1 500 μL Unconjugated Concentrated 200μg/mL
MS144B0 100 μL Biotin Concentrated 200μg/mL
MS144B1 500 μL Biotin Concentrated 200μg/mL
MS144B 1 mL Biotin Concentrated 200μg/mL
MS144P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS144PABX 200 μL Unconjugated Concentrated 1mg/mL
MS144P 1 mL Unconjugated Concentrated 200μg/mL
MS144R1 1 mL Unconjugated Prediluted -
MS144R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS144P0 Supplier Epredia Supplier No. MS144P0

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-1, Mouse Monoclonal Antibody.

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: VU-1D9

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma cells

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VU-1D9
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Small cell lung carcinoma cells
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.