Learn More
Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: VU-1D9
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Small cell lung carcinoma cells
Molecular Weight: 40-43 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Epithelial Specific Antigen/Ep-CAM Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | VU-1D9 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Small cell lung carcinoma cells |
| Quantity | 7 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.