Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epithelium Endothelial Cells Mouse anti-Human, Unconjugated, Clone: 18.29 (PHM-5), MilliporeSigma™

Catalog No. MAB430I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB430-I-100 100 μg
MAB430-I-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB430-I-100 Supplier MilliporeSigma Supplier No. MAB430I100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Epithelium Endothelial Cells, clone PHM-5, Cat. No. MAB430-I, is a mouse monoclonal antibody that detects human human epithelium/endothelial cells and has been tested for use in Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epithelium Endothelial Cells
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 18.29 (PHM-5)
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Glomerular endothelial cells;Glomerular epithelial cells
Host Species Mouse
Immunogen Human Glomerular cells.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001018121
Test Specificity Clone PHM-5 is a mouse monoclonal antibody that detects human epithelium/endothelial cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.