Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPLIN Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002310APC
Description
EPLIN Polyclonal antibody specifically detects EPLIN in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
EPLIN | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3 | |
Maps to a region between residues 500 and 550 of human epithelial protein lost in neoplasm beta using the numbering given in entry NP_057441.1 (GeneID 51474). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51474 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction