Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPLIN Antibody (OTI1A3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271932AF350
Description
EPLIN Monoclonal specifically detects EPLIN in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EPLIN | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
LIMA1 | |
Recombinant protein fragment corresponding to amino acids 1-275 of human EPLIN (NP_057441) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A3 | |
Western Blot, Immunohistochemistry | |
1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3 | |
Mouse | |
0.1 mL | |
51474 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction