Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPLIN Monoclonal Antibody (OTI1A3)
GREENER_CHOICE

Catalog No. PIMA527016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527016 Supplier Invitrogen™ Supplier No. MA527016
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EPLIN Monoclonal Antibody for Western Blot, IHC (P)

EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPLIN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LIMA1
Gene Accession No. Q9UHB6
Gene Alias 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3
Gene Symbols LIMA1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-275 of LIMA1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51474
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.