Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPLIN Mouse anti-Human, Rat, Clone: 6E6E6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | EPLIN |
---|---|
Clone | 6E6E6 |
Concentration | 0.55 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6508
![]() ![]() |
Proteintech Group Inc
660711IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes both of isoforms of EPLN.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.Specifications
EPLIN | |
0.55 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EPLIN, LIM domain and actin binding 1, LIMA1, SREBP3 | |
LIMA1 | |
IgG1 | |
Protein G | |
Antibody |
6E6E6 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q9UHB6 | |
300228, 51474 | |
EPLIN Fusion Protein Ag13288 | |
Primary | |
-20°C | |
LIMA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title