Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPLIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554904 Supplier Invitrogen™ Supplier No. PA554904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISENSYSLDD LEIGPGQLSS STFDSEKNES RRNLELPRLS ETSIKDRMAK YQAAVSKQSS STNYTNELKA SGGEIKIHKM EQKENVPPGP EVCITHQEGE KISANENSLA VRSTPAEDDS RDS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPLIN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIMA1
Gene Accession No. Q9UHB6
Gene Alias 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3
Gene Symbols LIMA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPLIN. Recombinant protein control fragment (Product #RP-91594).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.