Learn More
Invitrogen™ EPLIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120129
Description
Positive test controls include: A-549. The target is usually found in the following locations: Cell junction, Cytoplasm, cytoskeleton, focal adhesion. Immunogen sequence: DEISKPEVPE DVDLDLKKLR RSSSLKERSR PFTVAASFQS TSVKSPKTVS PPIRKGWSMS EQSEESVGGR VAERKQVENA KASKKNGNVG KTTWQNKESK GETGKRSKEG HSLEMENENL VENGADSDED DNSFLKQQSP QEPKSLNWSS FVDNTFAEEF TTQNQKSQDV ELWEGEVVKE LSVEEQIKRN RYYDEDEDEE.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
| EPLIN | |
| Polyclonal | |
| Unconjugated | |
| LIMA1 | |
| 1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UHB6 | |
| LIMA1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 560-759 of human LIMA1 (NP_057441.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.