Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPLIN Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531567
Description
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications
EPLIN | |
Polyclonal | |
Unconjugated | |
LIMA1 | |
1110021C24Rik; D15Ertd366e; Epithelial protein lost in neoplasm; epithelial protein lost in neoplasm beta; eplin; LIM domain and actin binding 1; LIM domain and actin-binding protein 1; LIMA1; mEPLIN; PP624; SREBP3; sterol regulatory element binding protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51474 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UHB6 | |
LIMA1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human EPLIN. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction