Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    EPLIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 166391AP150UL
 
            Description
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.Specifications
| EPLIN | |
| Polyclonal | |
| Unconjugated | |
| LIMA1 | |
| EPLIN, LIM domain and actin binding 1, LIMA1, SREBP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 300228, 51474, 65970 | |
| -20°C | |
| Liquid | 
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ERG0, Q9UHB6 | |
| LIMA1 | |
| EPLIN Fusion Protein Ag9994 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                             
                             
                             
                             
                             
                            