Learn More
Tocris Bioscience™ Eplivanserin hemifumarate

Description
Specifications
Specifications
| Chemical Name or Material | (1Z,2E)-1-(2-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one O-[2-(dimethylamino)ethyl]oxime hemifumarate |
| CAS | 130580-02-8 |
| Synonym | SR-46349 hemifumarate, SR-46349B |
| Quantity | 10 mg |
| Target | 5-HT2A Receptor Antagonists |
| Molecular Formula | C19H21FN2O2.0.5C4H4O4 |
| Formula Weight | 386.42 |
| Purity | >98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.