Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Eplivanserin hemifumarate
GREENER_CHOICESDP

Catalog No. 495850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-585-0 50 mg
49-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-585-0 Supplier Tocris Bioscience™ Supplier No. 4958/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective 5-HT2A antagonist

Eplivanserin hemifumarate is a potent and selective 5-HT2A antagonist (IC50 values are 5.8 and 120 and >100 nM for 5-HT2A, 5-HT2B and 5-HT2C, respectively). Attenuates cocaine-induced hyperactivity. Increases dopamine (DA) release in rat medial prefrontal cortex (mPFC); potentiates haloperidol-induced DA release in the mPFC and nucleus accumbens.

Specifications

Chemical Name or Material (1Z,2E)-1-(2-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one O-[2-(dimethylamino)ethyl]oxime hemifumarate
CAS 130580-02-8
Synonym SR-46349 hemifumarate, SR-46349B
Quantity 50 mg
Target 5-HT2A Receptor Antagonists
Molecular Formula C19H21FN2O2.0.5C4H4O4
Formula Weight 386.42
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.