Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPM2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111418
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111418 100 μL
PIPA5111374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111418 Supplier Invitrogen™ Supplier No. PA5111418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATGHTNEMKH TTDFYFNIAG HQAMHYSRIL PNIWLGSCPR QVEHVTIKLK HELGITAVMN FQTEWDIVQN SSGCNRYPEP MTPDTMIKLY REEGLAYIWM PTPDMSTEGR VQMLP Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 95%.

The Epilepsy, progressive myoclonus type 2A protein (EPM2A) is a dual-specificity phosphatase that associates with polyribosomes. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. EPM2A interacts with a number of proteins known to be involved in glycogen metabolism and has been shown to have robust phosphatase activity against a phosphorylated complex carbohydrate, suggesting that EPM2A may be involved in the regulation of glycogen metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPM2A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPM2A
Gene Accession No. O95278
Gene Alias epilepsy, progressive myoclonic epilepsy, type 2 gene alpha; epilepsy, progressive myoclonus type 2, Lafora disease (laforin); epilepsy, progressive myoclonus type 2A; epilepsy, progressive myoclonus type 2A, Lafora disease (laforin); EPM2; EPM2A; EPM2A glucan phosphatase, laforin; EPM2A, laforin glucan phosphatase; glucan phosphatase; Glycogen phosphatase; Lafora PTPase; laforin; LAFPTPase; MELF; RP1-28C20.2; Tg(TcraK,TcrbK)TG-BFlv; TG-B
Gene Symbols EPM2A
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EPM2A. Recombinant protein control fragment (Product #RP-101738).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.