Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPM2A Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | EPM2A |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111374
|
Invitrogen™
PA5111374 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111418
|
Invitrogen™
PA5111418 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: ATGHTNEMKH TTDFYFNIAG HQAMHYSRIL PNIWLGSCPR QVEHVTIKLK HELGITAVMN FQTEWDIVQN SSGCNRYPEP MTPDTMIKLY REEGLAYIWM PTPDMSTEGR VQMLP.
The Epilepsy, progressive myoclonus type 2A protein (EPM2A) is a dual-specificity phosphatase that associates with polyribosomes. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. EPM2A interacts with a number of proteins known to be involved in glycogen metabolism and has been shown to have robust phosphatase activity against a phosphorylated complex carbohydrate, suggesting that EPM2A may be involved in the regulation of glycogen metabolism.Specifications
EPM2A | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95278 | |
7957 | |
Recombinant Protein corresonding to Human EPM2A. Recombinant protein control fragment (Product #RP-91068). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
EPM2A | |
epilepsy, progressive myoclonic epilepsy, type 2 gene alpha; epilepsy, progressive myoclonus type 2, Lafora disease (laforin); epilepsy, progressive myoclonus type 2A; epilepsy, progressive myoclonus type 2A, Lafora disease (laforin); EPM2; EPM2A; EPM2A glucan phosphatase, laforin; EPM2A, laforin glucan phosphatase; glucan phosphatase; Glycogen phosphatase; Lafora PTPase; laforin; LAFPTPase; MELF; RP1-28C20.2; Tg(TcraK,TcrbK)TG-BFlv; TG-B | |
EPM2A | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title