Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPM2AIP1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142488
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless.
Specifications
| EPM2AIP1 | |
| Polyclonal | |
| Unconjugated | |
| EPM2AIP1 | |
| A930003G21Rik; EPM2A (laforin) interacting protein 1; EPM2A interacting protein 1; EPM2A-interacting protein 1; EPM2AIP1; KIAA0766; Laforin-interacting protein; mKIAA0766; My007 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 9852 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q7L775 | |
| EPM2AIP1 | |
| Peptide with sequence C-TRNQHTLSQPLTDEH. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction