Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPN2 Monoclonal Antibody (OTI1E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504302

Description
Epsin2 plays a role in the formation of clathrin-coated invaginations and endocytosis. It binds to EPS15 (via the NPF repeat domain), as well as to AP-2 and clathrin (via the DPW repeat domain). The protein resides in the cytoplasm, in punctate structures throughout the cell, associated with clathrin-coated vesicles, and is particularly concentrated in the region of the Golgi complex. Highest expression is found in brain, with lower levels detected in lung and liver.Specifications
EPN2 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95208 | |
EPN2 | |
Full length human recombit protein of human EPN2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E4 | |
Unconjugated | |
EPN2 | |
EHB21 | |
Mouse | |
Affinity Chromatography | |
RUO | |
22905 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction