Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPN2 Monoclonal Antibody (OTI1E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504302S


Description
EPN2 Monoclonal antibody specifically detects EPN2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
EPN2 | |
Monoclonal | |
0.74 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95208 | |
Mouse | |
Affinity chromatography | |
RUO | |
22905 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E4 | |
Unconjugated | |
EPN2 | |
9530051D10Rik; AA536924; EH domain binding protein epsin 2; EHB21; EPN2; Eps15 binding protein; EPS-15-interacting protein 2; epsin 2; epsin-2; Ibp2; Intersectin-EH-binding protein 2; KIAA1065 | |
Full length human recombinant protein of human EPN2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction