Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPN2 Polyclonal Antibody

EPN2 Polyclonal Antibody
Supplier: Thermo Scientific PA5109689
Description
EPN2 Polyclonal Antibody for Western Blot
Specifications
| EPN2 | |
| Polyclonal | |
| Unconjugated | |
| EPN2 | |
| 9530051D10Rik; AA536924; EH domain binding protein epsin 2; EHB21; EPN2; Eps15 binding protein; EPS-15-interacting protein 2; epsin 2; epsin-2; Ibp2; Intersectin-EH-binding protein 2; KIAA1065 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13855, 22905, 60443 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O95208, Q8CHU3, Q9Z1Z3 | |
| EPN2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse Epn2 (XP_006532226.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction