Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPO Monoclonal Antibody (OTI5D8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812059


Description
EPO Monoclonal antibody specifically detects EPO in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
EPO | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P27699, Q03060, Q03061 | |
Mouse | |
Protein A/G | |
RUO | |
12916, 1390, 25620 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI5D8 | |
Unconjugated | |
CREM | |
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2 | |
Full length human recombinant protein of human CREM (NP_874388) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction