Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPO Rabbit anti-Human, Unconjugated, Clone: EPO/3793R, Abnova™

Catalog No. 89339501 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-501 Supplier Abnova Corporation Supplier No. RAB00545
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

EPO Monoclonal antibody specifically detects EPO in Human samples. It is validated for ELISA, IHC-P.

Specifications

Antigen EPO
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EPO/3793R
Conjugate Unconjugated
Description erythropoietin
Gene EPO
Gene Alias EP, MGC138142
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 28-162 of human EPO
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.