Learn More
Invitrogen™ EpoR Monoclonal Antibody (38407)

Description
In sandwich ELISAs, no cross-reactivity or interference with recombinant human (rh) Epo, recombinant mouse Epo R, or rhThrombopoietin R was observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | EpoR |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 38407 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | EPOR |
| Gene Accession No. | P19235 |
| Gene Alias | EPO R; EPOR; EPO-R; Erythropoietin receptor; pEpoR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.