Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ 24-Place Fixed-Angle Rotor with Aerosol-Tight Cover
Click to view available options
Capacity:
24 x 1.5/2 mL
Description
Includes
Lid
Supplied metal lid (13-688-199) provides secure containment of potentially hazardous aerosols from materials such as HIV and radioactive samples.
- 24 × 1.5 to 2.0mL capacity
- Maximum speed/RCF: 16,400rpm/25,000 x g
Order Info
With accessory adapters (sold separately), this rotor can also accommodate 0.5mL, 0.25/0.4mL, Microtainer™ and 0.2mL PCR tubes.

Specifications
Specifications
Type | Fixed Angle Rotor |
Capacity | 24 x 1.5/2 mL |
Max. RCF | 25,000 x g |
Max. Speed | 16,400 rpm |
For Use With (Equipment) | Eppendorf 5417C/5417R Microcentrifuge |
Angle | Fixed |
Includes | Basic or aerosol-tight lid |
Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction