Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE

Catalog No. E0030133374 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Skirt Style:
Semi-skirted
Unskirted
Unskirted, Divisible
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
PCR Clean and Free of Bacterial DNA (Non-sterile)
Packaging:
5 Bags x 4 Plates
5 Bags x 5 Plates
Individually Blister Wrapped
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Purity or Quality Grade Packaging
E0030133374 Colorless Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates
E0030129326 Colorless Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped
E0030129334 Blue Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped
E0030129610 Colorless Semi-skirted Forensic DNA Grade (Non-sterile) Individually Blister Wrapped
E0030133366 Colorless Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates
E0030133390 Blue Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates
E0030133404 Blue Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates
E951020320 Yellow Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
E951020346 Green Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
E951020362 Blue Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
E951020389 Red Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
E951020303 Colorless Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
14-380-056 Colorless Semi-skirted PCR Clean (Non-sterile) 5 Bags x 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. E0030133374 Supplier Eppendorf™ Supplier No. 0030133374
Add to Cart
Add to Cart

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color Colorless
Product Type PCR Plate
Skirt Style Unskirted, Divisible
Material Polypropylene, Polycarbonate
Volume (Metric) Well 250 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Ideal for extreme applications such as multiple heat sealing.
Max. RCF 2250 x g
Profile Regular
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Well Color Colorless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.