Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - PCR Clean
GSA_VAGREENER_CHOICE

Catalog No. p-4695875
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL

Ensures comfortable handling while preventing damage to gloves. Eppendorf; Combitips advanced™ PCR Pipet Tips meet the increased requirements of a modern laboratory.

Combitips advanced replaces Combitips plus. Functioning according to the positive displacement principle, these always dispense the correct desired volume, independent of the liquid density and its flow characteristics. Working with radioactive, aggressive, toxic or infectious liquids also becomes safer due to hermetically sealed piston preventing aerosol contamination. The new individual color coding facilitates quick identification of the desired tip.

  • Ideal for use with all Eppendorf Repeaters™
  • Nine sizes offer a maximum range of volumes with individual dispensing from 0.1 to 50 mL
  • Funnel geometry prevents damage to gloves and ensures comfortable handling
  • Elongated design allows users to empty all standard laboratory tubes completely
  • Allow for single-hand operation; easy-reach dispenser box conveniently dispenses through a chute
  • Perform automatic Combitip recognition and volume calculation
  • 3D design on the upper rim allows for easy recognition
  • Purity grade: PCR Clean Quality - portioning in 4 resealable bags (25 Tips per bag) ensures dust-free storage
  • Additional purity grades available: standard Eppendorf Quality or Biopur™ (Cat. Nos. 13-683-700 through -726)

Ideal for a range of applications, including ELISA, RIA, PCR set-up and general sample preparation or other serial pipetting tasks.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.2 mL
Sterility Non-sterile
Color Crystal
Autoclavable Not Autoclavable
Clean Claims PCR Clean (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Reclosable Bags x 25 Pk.)
Product Type Pipet Tips
Color Code Light Blue
Disposable Single-use
For Use With (Equipment) Eppendorf Multipette
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.