Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ epTIPS LoRetention Tips, Eppendorf quality, Reloads

Catalog No. 02717350 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
2 to 200 μL
50 to 1000 μL
Color:
Blue
Dark Gray
Light Gray
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
02-717-350 0.5 to 20 μL Light Gray
02-717-349 0.1 to 10 μL Dark Gray
02-717-351 2 to 200 μL Yellow
02-717-352 50 to 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-717-350 Supplier Eppendorf™ Supplier No. 022493020

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Increase reproducibility and and notably reduce the loss of expensive reagents

The surface of epT.I.P.S. LoRetention is achieved through an innovative treatment at the molecular level: the Pearl Effect Technology. The tips are uncoated and free from additives, thus nothing can bleed into the sample. Liquids roll off completely, so that almost no sample remains in the tip. In comparison, the adhesive layer of liquid in standard tips retains considerably more sample material. This unique material property guarantees maximum sample recovery, improved reproducibility and higher sensitivity.

  • Ultra hydrophobic and homogeneous surface for maximum reproducibility
  • Ultra-hydrophobic surface for minimum loss of sample
  • Air bubbles and foaming are significantly reduced during pipetting
  • Autoclavable with high chemical resistance
  • Shelf life is 5 years
  • Available as Eppendorf Quality purity grade
  • Universal tip fit

PCR and real-time PCR preparation of enzyme solutions and master mixes; Isolation, purification and denaturation of proteins; Restriction digestions and ligations; Pipetting DNA ladders during gel electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 20 μL
Color Light Gray
Autoclavable Autoclavable
For Use With (Application) PCR and real-time PCR preparation of enzyme solutions and master mixes, isolation, purification and denaturation of proteins, restriction digestions and ligations, pipetting DNA ladders during gel electrophoresis
Format Rack
Tip Style Low Retention
Product Type Pipet Tip Refills
Product Line epT.I.P.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.