Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE

Catalog No. E0030127870 Shop All Eppendorf North America Products
Change view
Click to view available options
Product Type:
PCR Film
PCR Foil
Storage Film
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
E0030127870 Storage Film
E0030127854 PCR Foil
E0030127838 PCR Film
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0030127870 Supplier Eppendorf™ Supplier No. 0030127870
Add to Cart
Add to Cart

Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination

  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
No. per Pack 100 Pk.
For Use With (Application) High-strength adhesive that forms tight seals to minimize sample evaporation
Product Type Storage Film
Certified RNase/DNase-free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.