Learn More
Eppendorf™ HeatSealer S100 Accessory, Plate Adapters
Catalog No. 05412614 Shop All Eppendorf North America Products
Change view
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
05-412-614 PCR Plate Adapter
05-412-612 High Profile Adapter
05-412-613 Low Profile Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-412-614 Supplier Eppendorf™ Supplier No. 5391070026
Add to Cart
Add to Cart
For use with HeatSealer S100
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty and Services
One year
TRUSTED_SUSTAINABILITY
Specifications
Wattage 320 W
For Use With (Equipment) HeatSealer S100
Sealing Temperature Range 170°C
Product Type PCR Plate Adapter