Learn More
Eppendorf™ HeatSealer S200 Accessory, Plate Adapters
Catalog No. 05412617 Shop All Eppendorf North America Products
Change view
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
05-412-617 PCR Plate Adapter
05-412-615 High Profile Adapter
05-412-616 Low Profile Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-412-617 Supplier Eppendorf™ Supplier No. 5392070038
Add to Cart
Add to Cart
For use with HeatSealer S200
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty and Services
Two years
TRUSTED_SUSTAINABILITY
Specifications
Wattage 350 W
For Use With (Equipment) HeatSealer S200
Sealing Temperature Range 125°C to 200°C in 1°C increments
Product Type PCR Plate Adapter