Learn More
Eppendorf™ Mastercycler™ Nexus Thermal Cycler
Offers reliability and ease of use along with low energy consumption
Supplier:  Eppendorf™ 6348000029
 View more versions of this product
Catalog No. E6348000029
Description
Description
Easy expansion to three units with a small footprint fits perfectly on any lab bench. The flexlid accommodates all types of consumables, from low volume all the way up to 0.5mL PCR tubes.
  • Flexlid concept allows use of all types of consumables with automatic height adjustment of the lid
  • Intuitive graphic programming
  • Small footprint
  • Universal block for ultimate flexibility
  • Connect up to two Mastercycler Nexus Eco to your existing unit
  • Available with 12-column SteadySlope gradient
  • Email notification to let you know the status of a run
  • Protocol library for ease of use
  • Booking schedule that acts as screensaver
Note:
The eco unit (Cat. No. E6330000021) is not a standalone unit.  The eco unit must be connected to either the nexus gradient or nexus nongradient unit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 x 0.2 mL, 71 x 0.5 mL
4 to 99°C
120 V
Nexus SX1e eco; Silver Block
3°C/s. Heating; 2°C/s. Cooling
±0.2°C
≤±0.4°C at 95°C
120 V, 50/60 Hz
50 to 60 Hz
Aluminum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.