Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Microcentrifuge Tubes

Catalog No. 0540734 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
05-407-34 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-407-34 Supplier Eppendorf™ Supplier No. 022379224

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Eppendorf Safe-Lock Tubes are the result of 50 years of continuous improvement and development. The original Safe-Lock tubes protect your sample with excellent centrifugation stability, feature perfect sealing properties, and reliably prevent sample evaporation. Trust in the original Eppendorf Safe-Lock Tubes, because your samples deserve nothing but the best.

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Guaranteed functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1.5 mL
Color Red
Material Polypropylene
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.