Learn More
Eppendorf™ Pipetter Carousel Stand
Holds Eppendorf Research Plus and Reference pipetters as well as all older Eppendorf models
Supplier:  Eppendorf™ 022444905
Catalog No. 05-403-04
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
  • Pipetters attach to the stand by the handle, not the nose cone, which eliminates potential cross-contamination from stand to pipetters
  • Stores all Research and Research Plus single-channel and multichannel pipetters in minimal space
  • Suitable for use with all reference pipetters without an adapter
Specifications
Specifications
Not Autoclavable
6
Free Standing
Polycarbonate
Research plus and Reference pipettes and all older models of Eppendorf pipettes.