Learn More
Eppendorf™ Reference™ 2 Fixed Volume Pipets- Trade-In Offer
Catalog No. 05412551 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
1 μL
2 μL
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
2 mL
2.5 mL
Color:
Blue
Gray
Red
Yellow
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
05-412-551 20 μL Yellow
05-412-547 5 μL Gray
05-412-545 1 μL Gray
05-412-546 2 μL Gray
05-412-549 10 μL Yellow
05-412-548 10 μL Gray
05-412-550 20 μL Gray
05-412-553 50 μL Yellow
05-412-552 25 μL Yellow
05-412-554 100 μL Yellow
05-412-555 200 μL Yellow
05-412-556 200 μL Blue
05-412-557 250 μL Blue
05-412-558 500 μL Blue
05-412-559 1000 μL Blue
05-412-560 2 mL Red
05-412-561 2.5 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 05-412-551 Supplier Eppendorf™ Supplier No. 2231302055
Add to Cart
Add to Cart
Available in trade-in offer at reduced price
Exchange any brand pipet with brand new Reference 2 pipet (1μL to 2.5mL) at low costs
  • One-button operation pipet offers fast and ergonomic handling with reduced operating effort and active aerosol reduction
  • Reduced repetitive strain injuries by not having to move your thumb sideways
  • Spring loaded tip cone improves user reproducibility and ergonomics by lowering tip attachment and ejection force
  • Secondary user adjustment supports easy adjustment of pipet for accurate pipetting without need for full calibration
  • Four digit volume display with magnifying window for easier volume identification, volume lock feature prevents accidental changes in volume
  • Clear haptic feedback is felt after blowout step which avoids accidental tip ejections
  • Embedded RFID chip contains all relevant pipet data and TrackIT software for simple identification and documentation
  • Reference 2 multichannel pipets offers high throughput option including spring-loaded nose cones which can be switched on/off optionally
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Serial number is printed on multiple components of pipet
  • Volume labeled pipet for quick identification of volume size/tip size
  • Round upper part of pipet makes it possible to work in every position
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipet robust
  • Quick connect lower part enables easy cleaning and maintenance of pipet
  • Fully autoclavable pipet with smooth surface and few rough edges
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Yellow
Type Fixed Volume Pipette
Volume (Metric) 20 μL