Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ S-4-104 Rotor Adapters

Click to view available options
Capacity:
1 x 250 mL Tubes
1 x 500 mL Bottle
14 x 15 mL Conical Tubes
14 x 5 mL Conical Tubes
20 x 5.5/12 mL Tubes
23 x 2.6/8 mL Tubes
23 x 7/17 mL Tubes
4 x 750 mL Wide-mouth Bottles
5 x 50 mL Conical Tubes
50 x 1.5/2 mL Tubes
7 x 50 mL Conical Tubes
Description
Includes
2 piece set
- For use with Centrifuge 5810/5810 R
- Holds up to 4 × 750mL round buckets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Type | Rotor Adapter |
Capacity | 4 x 750 mL Wide-mouth Bottles |
For Use With (Equipment) | S-4-104 rotor for 5810/5810R centrifuge |
No. per Pack | 2 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction