Learn More
Eppendorf™ Trade-in Xplorer™ Electronic Pipetters, Multichannel
Catalog No. 14286541 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
15 to 300 μL
5 to 100 μL
50 to 1200 μL
Number of Channels:
8
12
Color:
Gray
Green
Orange
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Color
14-286-541 15 to 300 μL 8 Orange
14-286-537 0.5 to 10 μL 8 Gray
14-286-538 0.5 to 10 μL 12 Gray
14-286-539 5 to 100 μL 8 Yellow
14-286-540 5 to 100 μL 12 Yellow
14-286-542 15 to 300 μL 12 Orange
14-286-543 50 to 1200 μL 8 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-286-541 Supplier Eppendorf™ Supplier No. 486100148
Only null left
Add to Cart
Add to Cart
Specially designed for high professional standards to provide optimal support.
Pipetters are based on the proven Eppendorf PhysioCare™ concept. The features set new standards for simplicity, precision and reproducibility, which mean no more delays due to complicated programming or inflexible processes. Precisely adjustable parameters, maximally reproducible results, fatigue-free work and full control over the pipetting processes are achieved.
  • Intuitive user interface
  • Pipetting modes: autodispensing, dispensing, pipetting, mixing, manual pipetting
  • Universal nose cone fits most pipette tips
  • Highly reproducible results
  • Spring engaged nose cone for enhanced tip attachment
  • Light weight design offers unsurpassed ergonomics
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Number of Channels 8
Product Line Trade-in Xplorer
Type Electronic Pipette
Volume (Metric) 15 to 300 μL
Finger Style Ergonomic