Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPPK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566869 Supplier Invitrogen™ Supplier No. PA566869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWELLFSEAIS SEQRAMLAQQ YQEGTLSVEK LAAKLSATLE QAAATARVTF SGLRDTVTPG ELLKAEIIDQ DLY Highest antigen sequence identity to the following orthologs - mouse 38%, rat 36%.

Cytoskeletal linker protein that connects to intermediate filaments and controls their reorganization in response to stress (PubMed:15671067, PubMed:27206504, PubMed:23398049). In response to mechanical stress like wound healing, is associated with the machinery for cellular motility by slowing down keratinocyte migration and proliferation and accelerating keratin bundling in proliferating keratinocytes thus contributing to tissue architecture (PubMed:27206504, PubMed:23398049). However in wound healing in corneal epithelium also positively regulates cell differentiation and proliferation and negatively regulates migration thereby controlling corneal epithelium morphogenesis and integrity. In response to cellular stress, plays a role in keratin filament reorganization, probably by protecting keratin filaments against disruption. During liver and pancreas injuries, plays a protective role by chaperoning disease-induced intermediate filament reorganization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPPK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPPK1
Gene Accession No. P58107
Gene Alias 450 kDa epidermal antigen; epidermal autoantigen 450K; EPIPL; EPIPL1; epiplakin; epiplakin 1; EPPK1
Gene Symbols EPPK1
Host Species Rabbit
Immunogen Recombinant Human EPPK1. Recombinant protein control fragment (Product #RP-107520).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83481
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.