Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPPS, 99%

Catalog No. aaa1371414
Click to view available options
Quantity:
25 g
100 g

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol

EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Soluble in water. 25.2 g/L (20°C)

Notes
Store at room temperature. Incompatible with oxidizing agents. Store away from extreme heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
MDL Number MFCD00006160
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym hepps, epps, 4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, 1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl, n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid, 4-2-hydroxyethyl piperazine-1-propanesulfonic acid, 4-2-hydroxyethyl-1-piperazinepropane sulfonic acid, 4-2-hydroxyethyl-1-piperazinepropanesulphonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Melting Point 242°C (decomposition)
Quantity 25 g
Beilstein 3957385
Solubility Information Soluble in water. 25.2g/L (20°C)
Formula Weight 252.33
Percent Purity 99%
Chemical Name or Material EPPS
Hazard Category H302
Precautionary Statement P264-P270-P301+P312-P330-P501c
EINECSNumber 240-198-8
RTECSNumber TM2649130
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.